Learn More
Draeger Short Term Tubes
Click to view available options
Components Measured:
Mercaptan 0.1/a
i-Propanol 50/a
Unit Size:
Pack of 10
Description
- Quick and targeted measurements
- Easy handling either manually using the Draeger-Tube pump accuro or automatically with the Draeger X-act 5000
- Field of application includes air examinations at the workplace, technical gas analysis in areas of emission/immission concentrations, measurement of compressed air/gases Measures a large number of different gases and vapours
Specifications
Specifications
| Components Measured | i-Propanol 50/a |
| Type | Short-term Draeger Sampling Tubes |
| For Use With (Application) | Spot Measurement of More That 500 Different Gases |