Learn More
Nickel Electro™ Water Bath Drain Tap
Nickel Electro™ Water Bath Drain Tab is designed to fit in all Clifton™ bath drain outlets.
Brand:  Nickel Electro™ BX0688
Product Code. 12769735
Description
Description
  • Permits easy emptying of water baths
  • Providing a gated on/off valve to empty fluids conveniently from the bath
  • Nozzle supplied with tap for laboratory tubing
  • Before fitting make sure bath is empty of liquid. Unscrew the stop plug with a 6mm Allen key whilst holding the nut with a 21mm spanner or nut holder supplied with tap. Screw the Tap into position, recommend pinch tight.
Specifications
Specifications
Water Baths with Serial Numbers Up to 75100
Drain Tap