Learn More
asecos™ Drum Rack With Sump Pallet Steel
Product Code. 17806426 Shop All asecos Products
Click to view available options
Weight (Metric):
66 kg
73 kg
118 kg
139 kg
Unit Size:
Each
Product Code. 17806426
Brand: asecos™ 13246
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Drum Rack With Sump Pallet Steel provides safe storage and filling storage of substances hazardous to water of all hazard classes.
  • For the storage of oil, paint and lacquer.
  • Safe dispensing due to overhanging collecting sump.
Specifications
Capacity (English) 54.2 gal.
Capacity (Metric) 205 L
Material Steel
Type Drum Rack
Length (English) 52.36 in.
Length (Metric) 1330.00 mm
Width (English) 52.36 in.
Width (Metric) 1330.00 mm
Height (English) 61.81 in.
Height (Metric) 1570.00 mm
Depth (English) 32.09 in.
Depth (Metric) 815.00 mm
Weight (English) 306.44 lb.
Weight (Metric) 139 kg