Learn More
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Click to view available options
Capacity (Metric):
0.5 μL
1 μL
25 μL
50 μL
Unit Size:
Pack of 100
Length (Metric):
32 mm
65 mm
100 mm
Description
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Specifications
Specifications
| Capacity (Metric) | 1 μL |
| Quantity | 100/Pk. |
| Type | Capillary Tube |
| For Use With (Equipment) | For TLC Sample Applicators |
| Includes | 1 glass bulb holder assembly |