Learn More
KGW Isotherm™ DSS Stainless Steel Dewar Flasks
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
Height (Metric):
178 mm
190 mm
225 mm
305 mm
Unit Size:
Each
Description
Lid is sold separately; see KGW Isotherm Lids (10166941).
Specifications
Specifications
| For Use With (Application) | Used to Store and Transport liquid nitrogen ( LN2 ), Dry Ice ( CO2 ) and Other Coolants |
| Height (Metric) | 178 mm |
| Capacity (Metric) | 1 L |
| Diameter (Metric) Inner | 100 mm |
| Material | DSS Stainless Steel |