missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-B Instrument™ Durac™ 10/30 Ground Joint Liquid-In-Glass Thermometers, Organic Liquid Fill
Ground joint thermometers feature a 10/30 ground joint and are designed for use with chemistry glassware.
Brand: H-B Instrument™ B60804-1200
Description
- Manufactured for specific immersion depths
- Contain red organic liquid against white back glass
Specifications
Partial Immersion | |
Red Organic Liquid | |
-10°C to +250°C | |
1°C |
300 mm | |
12 in. | |
±1°C; ±1.5°C above 150°C | |
25 mm (1 in.) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- DURAC Liquid Fill for Thermometers
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder, or water spray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : acetone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction