Learn More
DWK Life Sciences DURAN™ Adapter, standard ground joint
The transition adapter connects two glass components with different standard ground joints.
20.55 EUR - 30.50 EUR
Product Code | Brand | Type | Joint Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Joint Size | Price | Quantity & Availability | ||||
10788251
|
DWK Life Sciences
241142404 |
Expansion | Socket NS 14/23, Cone NS 29/32 |
23.00 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
10499722
|
DWK Life Sciences
241142301 |
Expansion Joint, Non-DIN Size | Socket NS 14/23, Cone NS 24/29 |
23.70 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
10171401
|
DWK Life Sciences
241142901 |
Expansion Joint | Socket NS 19/26, Cone NS 29/32 |
22.70 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
10029840
|
DWK Life Sciences
241143203 |
Reduction Joint, Non-DIN Size | Socket NS 24/29, Cone NS 14/23 |
28.40 EUR
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
10622452
|
DWK Life Sciences
241143606 |
Expansion Joint, Non-DIN Size | Socket NS 24/29, Cone NS 29/32 |
20.55 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
10314961
|
DWK Life Sciences
241144208 |
Reduction | Socket NS 29/32, Cone NS 14/23 |
29.10 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
10182021
|
DWK Life Sciences
241142207 |
Expansion | Socket NS 14/23, Cone NS 19/26 |
22.35 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
10622642
|
DWK Life Sciences
241144302 |
Reduction | Socket NS 29/32, Cone NS 19/26 |
30.50 EUR
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
10757101
|
DWK Life Sciences
241142601 |
Reduction Joint | Socket NS 19/26, Cone NS 14/23 |
26.40 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
10203601
|
DWK Life Sciences
241142807 |
Expansion Joint, Non-DIN Size | Socket NS 19/26, Cone NS 24/29 |
22.75 EUR
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
10039840
|
DWK Life Sciences
241143306 |
Reduction Joint, Non-DIN Size | Socket NS 24/29, Cone NS 19/26 |
29.80 EUR
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
10151451
|
DWK Life Sciences
241144405 |
Reduction Joint, Non-DIN Size | Socket NS 29/32, Cone NS 24/29 |
30.50 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
Description
Transition adapters or reducers are glass components with two standard ground joints of different sizes. With the help of these adapters, glass components with different standard ground joints can be connected to each other.
- Tight closure due to standard ground joint
Specifications
1 Ea. |