missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Hecht™ DURAN™ Automatic Tilting Pipettes
Click to view available options
Capacity (Metric):
20 mL
Unit Size:
Each
Description
- Manufactured from DURAN™ borosilicate glass 3.3
- Without bottle
- Capacity of 20mL
- Capacity and joint size durably marked
Specifications
Specifications
| Capacity (Metric) | 20 mL |
| Material | Borosilicate Glass (3.3) |
| Standard Taper Joint | 29/32 |
| Product Type | Tilting Pipette |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction