Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Nitrogen Round Bottom Flasks
Product Code. 10294281
Click to view available options
Capacity (Metric):
24 mL
50 mL
100 mL
500 mL
1000 mL
Socket Joint:
Center Neck: 14/23 NS
Center Neck: 29/32 NS
Unit Size:
Each
Product Code. 10294281
Brand: Lenz Laborglasinstrumente™ 03360458
Only null left
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass, Duran Glass
Socket Joint Center Neck: 29/32 NS
Bottom Shape Round