Learn More
DWK Life Sciences DURAN™ Evaporating Flask, with standard ground joint, pear shape
Shop All DWK Life Sciences Products
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Description
Evaporating flasks are a variant of round flasks and have a pear-shaped shape. The standard ground joint allows a precise connection to other ground joint components, e.g. a rotary evaporator. Due to their shape, evaporating flasks allow a large uniform liquid surface, making them particularly suitable for the evaporation of solvents.
- Tight closure due to standard ground joint
- Fired-on, highly durable white print
- Pear shape
Specifications
Specifications
| Description | Autoclavable |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Evaporation of liquids; Heating; Suitable for heating baths and mantles; Mixing several components; Rotary evaporator flask, suitable for all standard rotary evaporators; Concentration of solutions |
| Quantity | 10/Pk. |
| Volume (Metric) | 500 mL |
| Diameter (Metric) | 101 mm |