missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Funnels with NS Cone

For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Funnels with NS Cone are manufactured from durable DURAN™ borosilicate glass.

Brand:  Lenz Laborglasinstrumente™ 05321004

Product Code. 10090211

  • 40.00 EUR / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Solid addition funnel with angle at 60°
  • Corrosion-free and resistant to thermal shock
  • With NS cone, featuring high taper accuracy and excellent tightness
Specifications

Specifications

80mm
Duran Glass, Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers