Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Screw Thread Tube
Duran™ Glass Screw Thread Tube with NS cone, cap, and sealing.
Brand: Lenz Laborglasinstrumente™ 01301814
This item is not returnable.
View return policy
Description
- Duran™ Glass
Specifications
NS 14/23 | |
Duran Glass | |
1 Ea. |
Screw-cap aperture, Silicone sealing ring, PTFE washers | |
Screw thread Tube | |
GL 18 |