Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable, hollow
Use as an alternative to magnetic stirrers with high viscosity or high stirring speeds.
Brand:  DWK Life Sciences 286306809
1021.25 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15894638
  • 1105.00 EUR / Pack of 50
Estimated Shipment: 16-01-2025
Description
Hollow DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
  • Hollow
  • Available in polished and ground version
  • The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
Stirring of reaction solutions with high viscosities or at high stirring speeds; For further processing to various stirrer shapes and types
Polished
WB 10