Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable, hollow
Use as an alternative to magnetic stirrers with high viscosity or high stirring speeds.
Brand:  DWK Life Sciences 286307008
807.50 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15901571
  • 850.00 EUR / Pack of 50
Estimated Shipment: 24-10-2024
Description
Hollow DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
  • Hollow
  • Available in polished and ground version
  • The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
Stirring of reaction solutions with high viscosities or at high stirring speeds; For further processing to various stirrer shapes and types
Ground Joint
WB 10