Learn More
DWK Life Sciences DURAN™ Petri Dish, pressed
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
90.90 EUR - 134.00 EUR
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Borosilicate Glass (3.3) |
For Use With (Application) | Cultivation on different culture media at the same time under the same conditions; Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; cell culture |
No. per Pack | 10/Pk. |
Product Code | Brand | Type | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Diameter (Metric) | Price | Quantity & Availability | ||||
11726958
|
DWK Life Sciences
217524801 |
Four-Sectional | 100 mm |
134.00 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
10132101
|
DWK Life Sciences
217504808 |
Half-Sectional | 100 mm |
134.00 EUR
Pack of 10 |
Estimated Shipment: 04-03-2025
Log in to see stock. |
||||
10142101
|
DWK Life Sciences
217534802 |
Three-Sectional | 100 mm |
134.00 EUR
Pack of 10 |
Estimated Shipment: 04-03-2025
Log in to see stock. |
||||
10796771
|
DWK Life Sciences
217544803 |
Without Sections | 100 x 20 mm |
104.00 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
15585016
|
DWK Life Sciences
217544606 |
Without Sections | 100 x 15 mm |
90.90 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
Description
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow. DURAN™ Petri dishes are manufactured from borosilicate glass 3.3 using a special manufacturing process. They allow an even distribution of the culture medium and guarantee a distortion-free view. The lids of the DURAN™ Petri dishes rest directly on the lower part and thus prevent turbulence of the protruding air space. This is well suited for reactions with long incubation times, at high temperatures or with sensitive culture media.
- Facilitated fractionated dilution smear by division (into 2, 3 or 4 parts)
- Available half-, three- and four-sectional or without sections
- Pressed
Specifications
Autoclavable | |
Borosilicate Glass (3.3) | |
Cultivation on different culture media at the same time under the same conditions; Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; cell culture | |
10/Pk. |