Learn More
DWK Life Sciences DURAN™ Powder Funnel, with short, wide stem
A powder funnel enables a clean transfer of solids and powders.
Brand:  DWK Life Sciences 213543307
131.10 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11716828
  • 142.00 EUR / Pack of 10
Estimated Shipment: 09-12-2024
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
  • With short and wide stem
  • Large stem diameter makes it suitable for transferring solids
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Specifications
55 mm
Autoclavable
10/Pk.
Borosilicate Glass (3.3)
Transferring of powdered substances and granulated material; Spillage of substances is prevented; Transferring