Learn More
DWK Life Sciences DURAN™ Spherical Joint, ball, polished
For grease-free flexible connections.
Brand:  DWK Life Sciences 286323608
177.65 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15834658
  • 192.00 EUR / Pack of 10
Estimated Shipment: 16-01-2025
Description
The non-sanded version of the DURAN™ spherical ground joint only seals over the sealing ring. Basically 2 versions of the sealing rings are available: - FEP-coated silicone rings - VITON sealing rings The nominal size is composed of the nominal diameter of the spherical ground joint and, if applicable, the clear width of the pipe diameter. This variant also enables flexible connections, these are in the angle range of 10-15°.
  • Flexible connections of up to 15° can be realised depending on the size of the cut (angle ≥ tightness reduced)
  • Grease-free working possible
  • A tight seal is made possible by the use of an additional sealing ring.
  • The nominal size describes the nominal diameter of the spherical ground joint.
  • Not self-supporting
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
For the production of grease-free, flexible connections (alignment in 360° possible); Installation in glass apparatus and devices
12 mm
2.2 mm