Learn More
DWK Life Sciences DURAN™ Tube with two Hose Connections
For attaching to a wide variety of apparatus.
Brand:  DWK Life Sciences 286023202
209.95 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15894508
  • 227.00 EUR / Pack of 250
Estimated Shipment: 16-01-2025
Description
Hoses can be connected to laboratory apparatus via DURAN™ tube nozzles. There is always a risk of injury due to glass breakage when dismantling tightly fitted hoses from glass olives. DWK therefore recommends the use of DURAN™ glass threads with plastic nozzles.
  • Hose olives enable the connection of hoses to laboratory apparatuses
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250/Pk.
For attaching to various apparatuses; Installation in different apparatus possible
Tube with Connections
11 mm