Learn More
Macherey-Nagel™ Durasil-25/UV254 TLC Plate
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 10 cm
5 x 20 cm
10 x 20 cm
20 x 20 cm
Quantity:
100 Pk.
200 Pk.
25 Pk.
50 Pk.
Unit Size:
Pack of 100
Pack of 200
Pack of 25
Pack of 50
Specifications
Specifications
| Material | Durasil-25 |
| Particle Size | 5 to 17 μm |
| Pore Size | 60 Å |
| Quantity | 50 Pk. |
| Type | TLC Plate |
| Fluorescent Indicator | UV254 |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.25 mm |
| Dimensions (L x W) | 5 x 10 cm |