Learn More
Applied Biosystems™ Dynabeads™ MX 12-tube Mixing Wheel
Dynabeads™ MX 12-tube Mixing Wheel
Brand:  Applied Biosystems™ 15903
484.50 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 10364393
  • 550.00 EUR / Each
Estimated Shipment: 03-01-2025
Description
Attachment for Dynabeads™ MX Mixer Base (Cat. No. 159-02). Designed to hold 12 sample tubes ranging from 1.5 ml (microcentrifuge) to 1.7 mm diameter. Requires use of Mixer Turret (Cat. No. 159-06) to affix to Mixer Base.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.5 mL
Mixing Wheel
Dynabeads™ Sample Mixers
12