Learn More
Abeba™ Dynamic 7315 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
68.55 EUR - 70.80 EUR
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | Black |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15632678
|
Abeba™
7315/35 |
35 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15642678
|
Abeba™
7315/36 |
36 |
68.55 EUR
1 pair |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
15652678
|
Abeba™
7315/37 |
37 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15662678
|
Abeba™
7315/38 |
38 |
68.55 EUR
1 pair |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
15672678
|
Abeba™
7315/39 |
39 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15682678
|
Abeba™
7315/40 |
40 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15692678
|
Abeba™
7315/41 |
41 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15602688
|
Abeba™
7315/42 |
42 |
70.80 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15612688
|
Abeba™
7315/43 |
43 |
68.55 EUR
1 pair |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
15622688
|
Abeba™
7315/44 |
44 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15632688
|
Abeba™
7315/45 |
45 |
68.55 EUR
1 pair |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
15642688
|
Abeba™
7315/46 |
46 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15652688
|
Abeba™
7315/47 |
47 |
68.55 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15662688
|
Abeba™
7315/48 |
48 |
68.55 EUR
1 pair |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather
- Black/brown
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Polyurethane |
Black | |
Slip-resistant | |
7315 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Abeba™ Dynamic 7315 Shoes