Learn More
Dynamin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-16244
Product Code. 18717503
  • 614.00 EUR / 0.10mL
Description
Dynamin 2 Polyclonal specifically detects Dynamin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
DNM2
98 kDa
RUO
IgG
CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII
Recombinant protein encompassing a sequence within the center region of human Dynamin 2. The exact sequence is proprietary.
0.1 mL
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Certificates
Promotions
Promotions
For Research Use Only