Learn More
Dynamin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16244
This item is not returnable.
View return policy
Description
Dynamin 2 Polyclonal specifically detects Dynamin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DNM2 | |
| 98 kDa | |
| RUO | |
| IgG |
| CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII | |
| Recombinant protein encompassing a sequence within the center region of human Dynamin 2. The exact sequence is proprietary. | |
| 0.1 mL | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |