missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysprosium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
Unit Size:
100mm
10mm
25mm
50mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
| Quantity | 50 mm |
| Odor | Odorless |
| Packaging | Mylar under Argon |
| Weight | ≈1.1g/mm |
| Form | Cast Rod |
| Percent Purity | 99.9% |
| Assay | (metals basis excluding Ta) |
| Chemical Name or Material | Dysprosium rod, 12.7mm (0.5 in.) dia. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction