Learn More
Invitrogen™ E-Gel™ Agarose Gels Starter Pak, 2% (EU only)
E-Gel™ Agarose Gels Starter Pak, 2% (EU only)
Brand:  Invitrogen™ G600002EU
Product Code. 10388982
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
E-Gel™ Single Comb Gels are supplied in a 12-well mini-gel format, with a choice of either SYBR Safe™, or ethidium bromide DNA stain. Each gel contains a single row of 12 sample wells. E-Gels are available in two pack sizes-a Starter Pak, and 18-gel multi-paks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E-Gel™ Agarose Gels Starter Pak, 2%, E-Gel™ Gel Type, Ethidium Bromide In-Gel Dye, 12 Wells, Single Comb Well Format, Store at Room Temperature, For Nucleic Acid Gel Electrophoresis and Blotting
Agarose Gels Starter Kit
6 E-Gels, the E-Gel™ PowerBase™ v.4, and a manual, 18-packs contain 18 gels and a manual
Room temperature
2%
DNA and RNA purification analysis, nucleic acid gel electrophoresis and blotting, agarose gel electrophoresis
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.