Learn More
Greiner Bio-One EASYseal™ Adhesive Microplate Sealer
For optical measurements and all laboratory purposes. Greiner Bio-One EASYseal™ Adhesive Microplate Sealer provides protection against evaporation and contamination.
Brand:  Greiner Bio-One 676001
Product Code. 15975331
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Coated with acrylate adhesive
  • Coverage/storage of microplates (e.g in ELISA applications)
  • Protection against evaporation and contamination
  • Suitable for optical measurements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent
100
Sealer
Microplate
Non-sterile
ELISA
Acrylate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.