missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Direct Human CTC Enrichment Kit
Click to view available options
Quantity:
100 mL Whole Blood
Unit Size:
1 set
Description
- The EasySep™ Direct Human CTC Enrichment Kit can be used to enrich Circulating Tumor Cells (CTCs) directly from human whole blood by immunomagnetic negative selection without the need for either density gradient centrifugation or RBC lysis
- Isolated cells are immediately ready for downstream analysis or assays
Specifications
Specifications
| Cell Type | Cancer Cells and Cell Lines |
| For Use With (Application) | Cell Isolation |
| Quantity | 100 mL Whole Blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction