Learn More
STEMCELL Technologies EasySep™ Human Basophil Isolation Kit
Immunomagnetic isolation of untouched human basophils
Brand: STEMCELL Technologies 17969
This item is not returnable.
View return policy
Description
- The EasySep™ Human Basophil Isolation Kit is designed to isolate basophils from a polymorphonuclear cell-rich fraction by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-basophils and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
- The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation of fresh peripheral blood
- This product replaces the EasySep™ Human Basophil Enrichment Kit (Catalog #19069) for even faster cell isolations
Specifications
Granulocytes and Subsets | |
Cell Isolation | |
1 x 109 Cells |