Learn More
STEMCELL Technologies EasySep™ Human Basophil Isolation Kit
Immunomagnetic isolation of untouched human basophils
Brand:  STEMCELL Technologies 17969
Encompass
Product Code. 15937025
  • 1015.50 EUR / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Description
  • The EasySep™ Human Basophil Isolation Kit is designed to isolate basophils from a polymorphonuclear cell-rich fraction by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-basophils and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation of fresh peripheral blood
  • This product replaces the EasySep™ Human Basophil Enrichment Kit (Catalog #19069) for even faster cell isolations
Specifications
Specifications
Granulocytes and Subsets
Cell Isolation
1 x 109 Cells