Learn More
STEMCELL Technologies EasySep™ Human CD4+ T Cell Enrichment Kit
Immunomagnetic isolation of untouched human CD4+ T cells
Brand: STEMCELL Technologies 19052
This item is not returnable.
View return policy
Description
- The EasySep™ Human CD4+ T Cell Enrichment Kit is designed to isolate CD4+ T cells from fresh or previously frozen peripheral mononuclear cells by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
T Cells, T Cells, CD4+ | |
Cell Isolation | |
1 x 109 cells |