Learn More
STEMCELL Technologies EasySep™ Non-Human Primate CD4+ T Cell Isolation Kit
Immunomagnetic isolation of untouched non-human primate CD4+ T cells
Brand:  STEMCELL Technologies 19582
Encompass
Product Code. 17117931
  • 1170.00 EUR / 1 set
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Description
  • The EasySep™ Non-Human Primate CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This kit has been verified for use with rhesus and cynomolgus macaques
Specifications
Specifications
T Cells, CD4+
Cell Isolation
1 x 109 cells