Learn More
STEMCELL Technologies EasySep™ Non-Human Primate CD4+ T Cell Isolation Kit
Immunomagnetic isolation of untouched non-human primate CD4+ T cells
Brand: STEMCELL Technologies 19582
This item is not returnable.
View return policy
Description
- The EasySep™ Non-Human Primate CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This kit has been verified for use with rhesus and cynomolgus macaques
Specifications
T Cells, CD4+ | |
Cell Isolation | |
1 x 109 cells |