Learn More
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
Click to view available options
Quantity:
100 μL
4 x 100 μL
Unit Size:
100µL
Pack of 4
Description
- A ready to use solution of PHA-L in aqueous buffer
- PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris
- It is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity
- This reagent is intended for use in in vitro activation of human and mouse leukocytes
Specifications
Specifications
| Quantity | 100 μL |
| Form | Liquid |
| Product Line | eBioscience |
| Product Type | Phytohemagglutinin-L (PHA-L) Solution |