Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 506 Conjugate
Product Code. 17104123
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. 17104123
Brand: Invitrogen™ 69431782
A streptavidin-fluorochrome conjugate for indirect staining protocols to detect biotinylated primary antibodies in flow cytometry
  • eBioscience Streptavidin eFluor 506 is a streptavidin-fluorochrome conjugate for indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. eFluor organic dyes are traditional fluorescent dyes for use in multiple applications, including multicolor flow cytometry and immunofluorescence microscopy.
  • To make fluorophore selection easier, all eFluor organic dyes are named after their emission wavelength, and they are fully compatible with most conventional dyes, fluorescent molecules, eBioscience buffers and fixatives, viability stains, and reagents.
  • The eFluor 506 organic dye is a violet-laser excitable fluorophore that has an emission peak of 506 nm. This dye is optimized for use in flow cytometry and, when combined with all five Super Bright dye antibody conjugates, can provide six detection channels of the violet laser.
  • We recommend using a 510/20 band pass filter or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.
  • Streptavidin eFluor 506 Conjugate has been tested by flow cytometric analysis to detect biotinylated primary antibodies. It can be used ≤ 0.125 μg per test. A test is defined as the amount (μg) of streptavidin fluorochrome conjugate that will stain a cell sample in a final volume of 100 μL.
  • Cell number should be determined empirically but can range from 105 to 108 cells/test. It is recommended that the streptavidin be carefully titrated for optimal performance in the assay of interest.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 0.2 mg/mL
Conjugate eFluor 506
Quantity 100 μg
Content And Storage Store at 2-8°C.
Shipping Condition Wet Ice
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye eFluor 506