missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDNRB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20964-1-AP-150UL
This item is not returnable.
View return policy
Description
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.Specifications
EDNRB | |
Polyclonal | |
Unconjugated | |
EDNRB | |
ABCDS, EDNRB, Endothelin B receptor, endothelin receptor type B, ET B, ET BR, ETB, ETBR, ETRB, HSCR, HSCR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13618, 1910 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24530, P48302 | |
EDNRB | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |