missing translation for 'onlineSavingsMsg'
Learn More
EDTA Disodium Salt, 0.1M/l Solution, Scharlab Product Code.: 11364104

EDTA Disodium Salt, 0.1M/l Solution, Scharlab

Product Code. 11364104
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11364104

Brand: Scharlab AC09701000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used in analytical chemistry, as a sequestering agent and for metals titration.

This volumetric solution was checked by means of potentiometric methods using Scharlau's calcium carbonate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).

  • Synonyms: Ethylenediaminetetraacetic acid disodium salt
  • Molecular Formula: C10H14N2Na2O8 • 2H20
  • Molar Mass: 372.24g/mol
  • CAS: 6381-92-6
  • EIN ECS No.: 205-358-3
  • Factor: 0.999–1.001
  • M=0.1 mol/l
  • N = 0.2

Specifications

CAS 6381-92-6
Density 1.01g/cm3
Molecular Formula C10H18N2Na2O10
MDL Number MFCD00150037,MFCD00003541
Quantity 1 L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Weight (g/mol) 372.24
PubChem CID 44120005
Formula Weight 372.24g/mo
Concentration or Composition (by Analyte or Components) 0.1 mol/L
Physical Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
EDTA Disodium Salt, 0.1M/l Solution, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.