missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15276-1-AP-20UL
This item is not returnable.
View return policy
Description
Necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA.Specifications
EIF1 | |
Polyclonal | |
Unconjugated | |
EIF1 | |
A121, EIF 1, EIF1, EIF1A, ISO1, SUI1, Sui1iso1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10209, 20918, 287703 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41567, P48024 | |
Eif1 | |
EIF1 Fusion Protein Ag7394 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |