missing translation for 'onlineSavingsMsg'
Learn More

ELAC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

193.30 EUR - 457.87 EUR

Specifications

Antigen ELAC2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18320697
View Documents Promotion Details
Bio-Techne
NBP3-04612-20UL
20 μL
193.30 EUR
20µL
Please to purchase this item. Need a web account? Register with us today!
18331384
View Documents Promotion Details
Bio-Techne
NBP3-04612-100UL
100 μL
457.87 EUR
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELAC2 Polyclonal specifically detects ELAC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

ELAC2
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
60528
IgG
Affinity purified
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Human
EC 3.1.26.11, elaC homolog 2 (E. coli), ElaC homolog protein 2, ELC2, FLJ10530, FLJ36693, FLJ42848, Heredity prostate cancer protein 2, HPC2elaC (E. coli) homolog 2, putative prostate cancer susceptibility protein HPC2/ELAC2, Ribonuclease Z 2, RNase Z 2, tRNA 3 endonuclease 2, tRNase Z (long form), tRNase Z 2, zinc phosphodiesterase ELAC protein 2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ELAC2 (NP_776065.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications