missing translation for 'onlineSavingsMsg'
Learn More

Elaidic acid, 98%, Thermo Scientific Chemicals

Store cold

48.94 EUR - 554.76 EUR

Chemical Identifiers

CAS 112-79-8
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00063954
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
PubChem CID 637517
ChEBI CHEBI:27997
IUPAC Name (E)-octadec-9-enoic acid
SMILES CCCCCCCCC=CCCCCCCCC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11402597
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14832.03
1 g
48.94 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
11412597
View Documents
Thermo Scientific Alfa Aesar
A14832.06
5 g
154.77 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
11422597
View Documents
Thermo Scientific Alfa Aesar
A14832.14
25 g
554.76 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

112-79-8
282.468
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
637517
(E)-octadec-9-enoic acid
C18H34O2
MFCD00063954
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
CHEBI:27997
CCCCCCCCC=CCCCCCCCC(=O)O

Specifications

112-79-8
288°C (100 mm)
98%
MFCD00063954
14,3533
Insoluble in water.
CCCCCCCCC=CCCCCCCCC(=O)O
282.468
CHEBI:27997
98%
41°C to 45°C
>110°C (230°F)
C18H34O2
1726543
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(E)-octadec-9-enoic acid
637517
282.47
Elaidic Acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 204-006-6

missing translation for 'rtecsNumber' : JX6125000

missing translation for 'tsca' : No

missing translation for 'storageNote' : Air sensitive

Recommended Storage : Keep cold; Store under Nitrogen

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Elaidic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.