Learn More
WTW™ Electrolyte for WTW™ SenTix™ ORP Electrode
Brand: WTW™ 109735
This item is not returnable.
View return policy
Description
Specifications
118°C | |
Clear | |
Combined SenTix™ ORP Electrode with Ag-Electrode | |
Electrolyte | |
-14°C | |
7 (at 25°C) |
250 mL | |
2 mol/L KNO3, 0.001 mol/L KCl | |
Electrolyte | |
2mol/L KNO3 0.001mol/L KCl, bottle with 250mL | |
Bottle | |
Yes |