Learn More
ELF4 Monoclonal Antibody (OTI1E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810143
Encompass_Preferred
 View more versions of this product
Product Code. 17523004
  • 540.87 EUR / 100µL
Description
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5′-WGGA-3′. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
Specifications
Specifications
ELF4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q99607
ELF4
Human recombit protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI1E11
Unconjugated
ELF4
ELFR, MEF
Mouse
Affinity Chromatography
RUO
2000
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions