Learn More
Merck PureProteome™ Carboxy FlexiBind Magnetic Bead System
For rapid and reproducible isolation of proteins
Brand:  Merck LSKMAG25CBX10
1685.82 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12601156
  • 1935.00 EUR / Each
Estimated Shipment: 15-01-2025
Description
Custom affinity bead generation for binding to target ligand.
  • Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Beads are immobilized in seconds
  • Increased kinetics of bead-protein binding enables shorter incubations
  • Significantly more affordable then competitive magnetic beads
Research Use Only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carboxy flexiBind Magnetic bead system
2.5μm
Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions
10mL
2° to 8°C
Magnetic Beads