Learn More
BD Emerald™ Three-Part Syringe
Product Code. 15691499
Click to view available options
Volume (Metric):
2 mL
3 mL
5 mL
10 mL
Graduations:
0.1 mL
0.2 mL
Unit Size:
Pack of 100
Pack of 2000
Product Code. 15691499
Brand: BD 307731
Combines optimum performance and reduced environmental impact. BD™ Emerald™ Three-Part Syringe is a general purpose syringe that contains up to 30% less material than other syringes to minimize waste. Syringes feature a center-mounted Luer connection and no needle.
  • Combines optimum performance and reduced environmental impact
  • Manufactured in a production plant powered by renewable energy
  • Crystal clear plastic allows for easy viewing of the contents
  • Green-colored piston crown and engraved shell pressure for precise and safe dosage
  • Robust piston stop prevents the rod from accidentally being knocked out
  • Rod designed for stable and comfortable grip during use
  • Thumb grip with the structure for secure grip during administration
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE marked, ISO 7886-1 and 549 compliant
Color Clear with Green Piston Crown
For Use With (Application) General purpose use and pressure applications
Product Type 3-part Syringe
Graduations 0.2 mL
Quantity 100/Pk.
Volume (Metric) 5 mL
Connection Type Centric Luer
Graduated Yes