missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ EnergyOne™ Barrel Pump Driven by Power Supply Unit

Buerkle™ EnergyOne™ Barrel Pump with Power Supply Unit Discharge Tube features a foil keyboard sealed off against dust, dirt and water.

Brand:  Buerkle™ 5900-2070

528.45 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 11971591

  • 723.00 EUR / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The barrel pump driven by power supply unit, EnergyOne, features a unique new hybrid system with complete flexibility and independence from battery charging.
  • Power supply via 230 V power supply unit, international plug adapter included with delivery
  • Optionally available with power supply unit or rechargeable battery (available as accessory)
  • Consisting of pump station, drive, power supply unit incl. connector and fixed discharge elbow
Specifications

Specifications

Unique hybrid power system, rechargeable battery and power supply unit are interchangeable
70 cm
Chemicals
240 V
CE Certified
Polypropylene
Energyone Barrel Pump Driven
Drive; Power Supply Unit; Pump Station, international plug adapter, connector and fixed discharge elbow
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.