Learn More
Tocris Bioscience™ Entecavir
Potent and selective hepatitis B virus inhibitor
Brand: Tocris Bioscience™ 6234/50
208.20 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Entecavir is a potent and selective hepatitis B virus reverse transcriptase inhibitor (EC50 = 3 nM); guanine analog. Inhibits hepatitis B virus replication in HepG2.2.15 cells. Displays selectively over HCMV, HSV-1, VZV, HIV and influenza (EC50 values are 15, 32, 30-60, >10 and >80 μM respectively). Orally bioavailable.Specifications
2-Amino-1,9-dihydro-9-[(1 S,3 R,4 S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-6 H-purin-6-one | |
142217-69-4 | |
277.28 | |
Store at -20°C | |
50 mg |
Inhibitor | |
C12H15N5O3 | |
BMS 200475, SQ 34676 | |
RNA/DNA Polymerase Inhibitors |
For Research Use Only