Learn More
Memmert™ Entry Port 23 mm for UF750plus, Custom Position TBC Left Side
Product Code. 17549605 Shop All Memmert Products
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
Product Code. 17549605
Brand: Memmert™ UF750plusF4
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in left side
Specifications
For Use With (Equipment) UF750plus Oven
Type Entry Port
Diameter (Metric) 23 mm