Learn More
Memmert™ Entry Port 38 mm for UF110
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF110F7
Product Code. 17578925
  • 221.00 EUR / Each
Estimated Shipment: 07-02-2025
Description
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
Specifications
UF110 Oven
Entry Port
38 mm