Learn More
Memmert™ Entry Port Left (Individual Location)
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Entry port flap closure custom TBC position in left side
- Can be closed by the flap
Specifications
Specifications
| Color | Clear |
| Description | Can be closed by flap |
| For Use With (Equipment) | IPP410eco |
| Diameter (Metric) | 23 mm |