Learn More
Memmert™ Entry Port Left (Individual Location)
Product Code. 17160155 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17160155
Brand: Memmert™ IPP2200ecoF4
Only null left
Add to basket
Add to basket
Entry port flap closure custom TBC position in left side
  • Entry port flap closure custom TBC position in left side
  • Can be closed by the flap
Specifications
Color Clear
Description Can be closed by flap
For Use With (Equipment) IPP2200eco
Diameter (Metric) 23 mm