Learn More
Memmert™ Entry Port Right Centre/Centre
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Standard position right centre/centre
- Not for model sizes 110 and 260 with light module
- Not for model size 110 with light module
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | HPP1060eco |
| Diameter (Metric) | 23 mm |