Learn More
Memmert™ Entry Port Standard Position
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Standard position rear (not in combination with illumination units)
- Moisture tight, can be closed by silicone stopper
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | HPP1060eco |
| Material | Silicone |
| Diameter (Metric) | 40 mm |