missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Eosin Methylene Blue Agar (EMB Agar)
Eosin Methylene Blue Agar (EMB Agar) for the isolation and enumeration of coliforms.
Brand: Scharlab 01-068-500
This item is not returnable.
View return policy
Description
- BAM/ISO/USP
- Selective differential medium
- For the isolation and enumeration of coliforms
- According to ISO 21150 and USP standard
Specifications
ISO 21150, USP | |
Eosin Methylene Blue Agar | |
Peptone 10.00 g/L, Lactose 10.00 g/L, Dipotassium Phosphate 2.00 g/L, Eosin Y 0.400 g/L, Methylene Blue 0.065 g/L, Agar 15.00 g/L | |
30°C to 35°C | |
6.90 ±0.2 |
Solid | |
Selective and Differential Medium | |
Isolation and Enumeration of Coliforms | |
24 to 48 hr. | |
500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction