missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelial Membrane Antigen/EMA Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia MS-348-P1
This item is not returnable.
View return policy
Description
Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown to be useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.Host Species: Mouse
Clone: GP1.4
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: DTRP
Immunogen: Human milk fat globule membranes
Molecular Weight: 265-400 kDa
Positive Control: Breast or Colon CA
Cellular Localization: Cytoplasmic and Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Epithelial Membrane Antigen/EMA Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Oncogenesis | |
Human | |
IgG1 |
Immunohistochemistry (Paraffin) | |
GP1.4 | |
Unconjugated | |
Human milk fat globule membranes | |
IVD | |
Primary | |
Concentrated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction